주요 문서
BAH420530002
SP Bel-Art® Dry-Keeper Small, Stacking Gas-Purge Desiccator Cabinet
polystyrene, volume 0.1 ft3
로그인조직 및 계약 가격 보기
About This Item
UNSPSC 코드:
41123002
NACRES:
NB.43
material
polystyrene
포장
pkg of 1 ea
제조업체/상표
Bel-Art H42053-0002
용적
0.1 ft3
일반 설명
Small, clear polystyrene desiccator cabinet safely stacks up to 5 units high for maximum use of lab bench space.
  • Pull down door slides out of the way into a compartment beneath the cabinet
  • Rear mounted gas ports are tapered to accept tubing with ¼ to ⅜” I.D.
  • Interior volume: 0.1 cu. ft
  • Gas ports can be closed to use desiccator with standard desiccant cartridges
법적 정보
Bel-Art is a registered trademark of SP Scienceware, a division of SP
가장 최신 버전 중 하나를 선택하세요:
시험 성적서(COA)