주요 문서
BAF379440000
SP Bel-Art® Sterileware Plastic Mini Tongs
polystyrene, L 4.25 in. (11 cm), pkg of 25 ea (individually wrapped)
로그인조직 및 계약 가격 보기
About This Item
UNSPSC 코드:
41122800
NACRES:
NB.35
material
polystyrene
포장
pkg of 25 ea (individually wrapped)
제조업체/상표
Bel-Art F37944-0000
길이
4.25 in. (11 cm)
일반 설명
These small, lightweight tongs are ideal for sampling small quantities of material ranging from food to pharmaceuticals.
특징 및 장점
  • Ridged jaws securely hold wet or dry samples
  • One piece, sterile tongs are made from polystyrene plastic
  • Individually packaged, sterilized by gamma irradiation, lot stamped for accurate lot tracing
법적 정보
Bel-Art is a registered trademark of SP Scienceware, a division of SP
Sterileware is a trademark of SP Scienceware, a division of SP
가장 최신 버전 중 하나를 선택하세요:
시험 성적서(COA)