MABF3126
Anti-DR4/CD261 Antibody, clone DJR1
Synonym(s):
Death receptor 4, TNF-related apoptosis-inducing ligand receptor 1, TRAIL receptor 1, TRAIL-R1, Tumor necrosis factor receptor superfamily member 10A
biological source
mouse
Quality Level
antibody form
purified antibody
antibody product type
primary antibodies
clone
DJR1, monoclonal
mol wt
calculated mol wt 50.09 kDa
purified by
using protein G
species reactivity
human
packaging
antibody small pack of 100 μg
technique(s)
flow cytometry: suitable
isotype
IgG1κ
epitope sequence
Extracellular domain
Protein ID accession no.
UniProt accession no.
storage temp.
2-8°C
Gene Information
human ... TNFRSF10A(8797)
General description
Specificity
Immunogen
Application
Evaluated by Flow Cytometry in BHK cells transfected with Human DR4.
Flow Cytometry Analysis: 1.0 µg of this antibody detected DR4/CD261 in BHK cells transfected with Human DR4.
Tested applications
Flow Cytometry: A representative lot detected DR4/CD261 in Flow Cytometry applications (Uno, K., et al. (2003). Blood. 101(9); 3658-67; Lee, H., et al. (2019). Cancer Res. 79(11); 2839-2852).
Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.
Physical form
Storage and Stability
Other Notes
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service