MilliporeSigma
A novel benzenediamine derivate rescued mice from experimental sepsis by attenuating proinflammatory mediators via IRAK4.
American journal of respiratory cell and molecular biology (2014-03-05)
Huan Dou, Yuxian Song, Xianqin Liu, Liu Yang, Nan Jiang, Dai Chen, Erguang Li, Renxiang Tan, Yayi Hou
RÉSUMÉ
We designed and synthesized a novel benzenediamine derivate, FC-99, that was tested for its ability to protect mice from experimental sepsis. Moreover, we sought to determine whether FC-99 could control a bacterial infection and to clarify the mechanism by which FC-99 inhibited LPS-activated macrophages. The effects of FC-99 on inflammation were evaluated in two experimental sepsis models and in cultured macrophages. Microarrays and docking and molecular dynamics simulations were used to determine the target of FC-99. Surface plasmon resonance and molecular detection were performed to confirm the direct interaction of FC-99 with its target. FC-99 protected mice from experimental sepsis. The mice that received FC-99 exhibited a diminished inflammatory response, had a lower local bacterial burden, and experienced a significantly improved survival rate. Genome-wide transcriptional profiling of FC-99-treated macrophages identified IRAK4 as a drug-regulated gene involved in LPS/TLR4 signaling. A computer search and calculations indicated that IRAK4 directly interacted with FC-99. Surface plasmon resonance, IRAK4-regulated signaling pathway analysis, and gene expression profiling of proinflammatory mediators confirmed the direct interaction between FC-99 and IRAK4. FC-99 is a potential therapeutic molecule for sepsis that alleviated experimental sepsis by directly inhibiting IRAK4 activation, which represents a novel target for sepsis therapy.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
FC-99 hydrochloride, ≥98% (HPLC)