Principaux documents
58249
Raccords unions Valco®
internal reducing, 1/4 to 1/16 in., bore size 0.75 mm
Synonyme(s) :
UNION REDUCTEUR Valco® 250-062
About This Item
Code UNSPSC :
41115700
Matériaux
stainless steel
Description
zero dead volume 0.75 mm bore
Conditionnement
pkg of 1 ea
Taille de l'alésage
0.75 mm
Raccords
1/16 in. Valco (nut and ferrule included)
1/4 in. Valco (nut and ferrule included)
Compatibilité
1/4 to 1/16 in.
Description générale
Acier inoxydable, sauf indication contraire. Écrous et ferrules inclus.
Produits recommandés
Discover LiChropur reagents ideal for HPLC or LC-MS analysis
Informations légales
Valco is a registered trademark of Valco Instruments Co. Inc.