Principaux documents
57624
Boucle d′injection pour injecteur modèle 7725
volume 10 μL
About This Item
Code UNSPSC :
41115854
Volume
10 μL
Description générale
Sample loop for model 7725 injector (volume 10 μL) is designed for advanced fluid-handling solutions for a broad range of analytical instrumentation and in vitro diagnostic systems. It finds application in high performance liquid chromatographic systems and is quite accurate and precise in injecting 1 μL-5 mL concentration of samples.
Produits recommandés
Discover LiChropur reagents ideal for HPLC or LC-MS analysis