Principaux documents
20352
Extracteur de septum
hook design
About This Item
Code UNSPSC :
47131900
Conditionnement
pkg of 1 ea
Technique(s)
gas chromatography (GC): suitable
Longueur
4 in.
Description générale
The hook septum puller is great for removing soft silicone septa from injection ports, and graphite ferrules from columns nuts. It has dozens of other uses around the lab.