Accéder au contenu
MilliporeSigma
Toutes les photos(4)

Principaux documents

ZRB2910

Anti-GFRAL Antibody, clone 1F5 ZooMAb® Rabbit Monoclonal

newenhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonyme(s) :

C6orf144, UNQ9356/PRO34128

Se connecterpour consulter vos tarifs contractuels et ceux de votre entreprise/organisme


About This Item

Code UNSPSC :
12352203

Source biologique

rabbit

Niveau de qualité

Produit recombinant

expressed in HEK 293 cells

Conjugué

unconjugated

Forme d'anticorps

purified antibody

Type de produit anticorps

primary antibodies

Clone

1F5, recombinant monoclonal

Description

recombinant, expressed in HEK 293 cells

Gamme de produits

ZooMAb® learn more

Forme

lyophilized

Poids mol.

calculated mol wt 44.5 kDa
observed mol wt ~56 kDa

Produit purifié par

using Protein A

Espèces réactives

human

Conditionnement

antibody small pack of 25

Caractéristiques du produit alternatif plus écologique

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

Validation améliorée

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

Technique(s)

affinity binding assay: suitable
flow cytometry: suitable
immunocytochemistry: suitable
western blot: suitable

Isotype

IgG

Séquence de l'épitope

N-terminal extracellular domain

Numéro d'accès Protein ID

Numéro d'accès UniProt

Autre catégorie plus écologique

Conditions d'expédition

ambient

Température de stockage

2-8°C

Informations sur le gène

human ... GFRAL(389400)

Catégories apparentées

Description générale

We are committed to bringing you greener alternative products, which adhere to one or more of the 12 Principles of Green Chemistry. This antibody is preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed, and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies. Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Spécificité

Clone 1F5 is a ZooMAb® rabbit recombinant monoclonal antibody that detects GFRAL. It targets an epitope within 25 amino acids from the N-terminal extracellular domain.

Immunogène

KLH-conjugated linear peptide corresponding to 25 amino acids from the N-terminal extracellular domain of human GFRAL.

Application

Quality Control Testing

Evaluated by Western Blotting in Neuro-2a cell lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected GFRAL in Neuro-2a cell lysate.

Tested Applications

Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected GFRAL in lysates from A-172 cells and MDA-MB-435 cells.

Flow Cytometry Analysis: 0.1 µg from a representative lot detected GFRAL in one million MDA-MB-435 cells.

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected GFRAL in HEK-293 cells.

Affinity Binding Assay: A representative lot of this antibody bound with GFRAL peptide with at least one hundred thousand-fold (100,000X) higher than with non-specific control peptide.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Description de la cible

GDNF family receptor alpha-like (UniProt: Q6UXV0) is encoded by the GFRAL (also known as C6orf144, UNQ9356/PRO34128) gene (Gene ID: 389400) in human. GFRAL is a glycosylphosphatidylinositol (GPI)-anchored protein, a member of the glial cell line derived neurotrophic factor (GDNF) receptor family. GFRAL is the receptor for Growth Differentiation Factor 15 (GDF15). The GFRAL-GDF15 axis plays a crucial role in regulating energy homeostasis and body wight. GFRAL has been studied in the context of obesity, where its signaling pathway might be targeted to suppress appetite and reduce food intake. Additionally, GFRAL-GDF15 signaling has been linked to cancer-related cachexia, a syndrome characterized by weight loss and muscle wasting. This ZooMAb® recombinant monoclonal antibody, generated by our proprietary technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Suriben, R., et al., (2020), Nature Medicine, 26, 1264 1270; Sabatini, P.V., et al., (2001), PNAS, 118:8, e2021357118)

Forme physique

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS with 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 µL.

Reconstitution

300 µg/mL after reconstitution at 25 µL per vial. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Stockage et stabilité

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 µL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Informations légales

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Clause de non-responsabilité

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Vous ne trouvez pas le bon produit ?  

Essayez notre Outil de sélection de produits.


Faites votre choix parmi les versions les plus récentes :

Certificats d'analyse (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

Si vous avez besoin d'assistance, veuillez contacter Service Clients

Déjà en possession de ce produit ?

Retrouvez la documentation relative aux produits que vous avez récemment achetés dans la Bibliothèque de documents.

Consulter la Bibliothèque de documents

Notre équipe de scientifiques dispose d'une expérience dans tous les secteurs de la recherche, notamment en sciences de la vie, science des matériaux, synthèse chimique, chromatographie, analyse et dans de nombreux autres domaines..

Contacter notre Service technique