Principaux documents
Z769363
Branson CPX series ultrasonic bath with digital timer
bath volume 1.9 L, AC/DC input 120 V AC
Synonyme(s) :
ultrasonic bath sonicator, ultrasonic cleaning bath
About This Item
Code UNSPSC :
41103706
Caractéristiques
model CPX1800
Conditionnement
pack of 1 ea
Fabricant/nom de marque
Branson CPX-952-119R
Taille du bain
150 mm (6 in.) × 140 mm (5.5 in.) × 100 mm (4 in.)
Volume du bain
1.9 L
Description générale
With the Bransonic® CPX, we′ve gone digital and defined a new category in ultrasonic baths. We′ve reimagined our iconic line of ultrasonic baths for total control with a fully digital Tower Control Panel. The CPX also features our new self-adaptive technology that actively tracks and adapts to changes within the bath.
All Bransonic® baths can also be used for cell separation, sample preparation and degassing of liquids.
Note: Parts should not be allowed to rest on tank bottom, see accessories for options.
Caractéristiques et avantages
  • Degassing - can be set up to 99 minutes
  • Timer - can be set to 99 minutes or run continuously
  • Self-Adaptive technology
  • Digital tower control panel
  • Sleep mode
  • All 220-230 V units meet CE standards
Informations légales
Bransonic is a registered trademark of Branson Ultrasonics Corp.