Principaux documents
Z765600
Tube rack for Benchmixer XL multi-tube vortexer
Holds 15 x 25 mL tubes
About This Item
Code UNSPSC :
41122804
Nomenclature NACRES :
NB.77
Caractéristiques
Holds 15 x 25 mL tubes
Conditionnement
pkg of 1 ea
Fabricant/nom de marque
Benchmark Scientific BV1010-25
Description générale
Benchmixer XL vortexer head adapters to hold multiple tubes.
This rack enables the use of 5 x 25 mL tubes in the BenchMixer XL heavy-duty hands-free mixing vortex mixer. It is ideal for the QuEChERS pesticide analysis method.
Informations légales
Benchmixer is a trademark of Benchmark Scientific, Inc.