Principaux documents
Z760978
Cap inserts for cryotubes
pack of 200 ea
About This Item
Code UNSPSC :
41121704
Nomenclature NACRES :
NB.22
Caractéristiques
(1 bag of each colour: blue, yellow, green, purple, red, white)
Conditionnement
pack of 200 ea
Fabricant/nom de marque
TPP 99020
Catégories apparentées
Description générale
Color-coding inserts in different colors facilitate the labeling and reliable identification of samples during cryogenic sample storage.