Principaux documents
Z744241
Wilmad® 4 mm Bruker® MAS Rotor Set
includes 2 Kel-F caps and 1 Torlon® cap
Synonyme(s) :
Solid State NMR, MAS NMR, Solid State NMR, MAS NMR,
About This Item
Code UNSPSC :
23152100
Nomenclature NACRES :
NB.15
Conditionnement
pack of 1 ea (2 Kel-F caps and bottom plugs)
Fabricant/nom de marque
Wilmad WP-501-4180-SET-1
Description générale
Manufactured from the highest purity Zirconia, MAS Rotors provide the NMR spectroscopist with the ultimate alternative for analysis of solid samples; the need to solvate is eliminated. They are available for most present solid state NMR spectrometers. MAS Rotors are carefully examined for material irregularities by optical methods before and after the precision machining process. Without over-spinning, each rotor is spin tested to only the highest specified spinning speed. Overspin testing, a common practice with some rotor brands, can actually damage the rotor prior to use. High precision (which becomes a necessity for proper spinning performance) is maintained in the manufacture of the end caps. Most caps are fitted with O-rings for better sealing; some have axial holes for venting. Advantages of Zirconia Rotors Most homogeneous ceramic material structure. Zirconia Rotor Body has higher strength (1,000 MPa, greater than Si3N4). Highest spinning speed, high precision superior finish (0.05 µ mirror gloss). Some caps have a uniquely designed detachable O-ring for use when a gas-tight environment is required for the sample.
Informations légales
Bruker is a registered trademark of Bruker-Physik AG
Torlon is a registered trademark of Amoco Chemicals Corporation
Vespel is a trademark of E. I. du Pont de Nemours and Company
Wilmad is a registered trademark of SP Industries