Principaux documents
Z741770
Sigma-Aldrich
Sigma Aldrich® Dual Run & Blot Accessory
Cooling Block
About This Item
Code UNSPSC :
41100000
Nomenclature NACRES :
NC.01
Description générale
Sigma Aldrich gels run best on our Dual Run and Blot unit. This unique apparatus permits the running of both 10x10cm and 8x10cm gels. The unit is available in dual and quad form enabling up to 4 gels to be run at one time.
Both units can also be used to complete Western blots. The unique electrode arrangement provides a gradient electric field which ensures that both small and large proteins migrate from the gel at a comparable rate.
Informations légales
SIGMA-ALDRICH is a registered trademark of Merck KGaA, Darmstadt, Germany