Principaux documents
Z740363
Eppendorf® Reference® 2 Variable Volume Pipettor
volume -0.5-10 μL, 10-100 μL, 30-300 μL, 100-1000 μL, 0.5-5 mL, 1-10 mL, pack of 6 ea
Synonyme(s) :
Adjustable volume pipette
About This Item
Code UNSPSC :
41121500
Nomenclature NACRES :
NB.01
Description
with stand
Caractéristiques
electronic: no
single-channel
Conditionnement
pack of 6 ea
Fabricant/nom de marque
Eppendorf® 2231302009
Plage de mesure du volume
0.5-10 μL
0.5-5 mL
1-10 mL
10-100 μL
100-1000 μL
30-300 μL
Volume
1000 μL
Description générale
The Reference 2 is the successor to the highly successful Reference pipette, and enhances the range′s already high standards with a new design, reduced weight and operating forces and multi-channel versions. As the premium product in Eppendorf′s pipette portfolio, the Reference 2 provides the most accurate results possible alongside robust, reliable handling and optimum user safety.
These levels of security make the Reference 2 ideal for use with precious liquids, or in any application requiring extremely high levels of accuracy. When there can be no compromise on quality and reproducability, Eppendorf′s new Reference 2 is an ideal instrument for these liquid handling applications.
Conditionnement
Includes: 6 adjustable-volume pipettes (0.5-10 μL, 10-100 μL, 30-300 μL, 100-1000 μL, 0.5-5 mL, 1-10 mL), 1 full box of Eppendorf pipette tips for each pipette volume (excludes 5 mL and 10 mL tips), includes a carousel stand
Informations légales
Eppendorf is a registered trademark of Eppendorf AG
Reference is a registered trademark of Eppendorf AG