Principaux documents
Z692786
microDOC Compact Gel Documentation System, with UV Transilluminator (UVT254/312)
AC/DC input 110 V AC
About This Item
Code UNSPSC :
41105300
Fabricant/nom de marque
Cleaver CSL-MDOCUV254/312$
Description générale
System comprises of:
Canon G12 digital
2MB storage card
Ethidium bromide & close up +3 filters
Lift-off hood with easy access door and built-in inner lights
8 in. TFT color monitor
12 x 12 cm UV 254/312 nm dual transilluminator
This option combines the microDOC Compact Gel Documentation System with a 12 x 12 cm UV 254/312 nm dual tranilluminator.
Available with optional TotalLab 1D software. Main software functions include: lane creation, background subtraction, band detection, profile deconvolution, and molecular size, pI and quantity calibration.
Informations légales
TotalLab is a trademark of Nonlinear Dynamics Ltd.