Principaux documents
Z351644
Sigma-Aldrich
Eppendorf® epT.I.P.S. Biopur®, singles
pack of 1000 ea (individually wrapped)
About This Item
Code UNSPSC :
41121600
Nomenclature NACRES :
NB.01
Matériaux
colorless
Description
PCR inhibitor free
Stérilité
sterile
Caractéristiques
ATP free
DNA free (Bacterial (E. Coli))
DNA free (Human)
DNase free
RNase free
barrier: no
pyrogen free
Conditionnement
pack of 1000 ea (individually wrapped)
Fabricant/nom de marque
Eppendorf® 022492225
Longueur
71 mm
Volume maximum
1 mL
Description générale
Biopur guaranteed sterile, pyrogen-free, RNase-free, DNA-free and ATP-free, for applications in pharmaceutical and food industries, molecular biology and cell technology.
Batch specific certificates available on the internet at http://www.eppendorf.com
Application
Eppendorf® epT.I.P.S.® Biopur®, singles are ideal for molecular biology, pharmaceutical, medical, and cell technology applications.
Informations légales
Biopur is a registered trademark of Eppendorf AG
Eppendorf is a registered trademark of Eppendorf AG
epT.I.P.S. is a registered trademark of Eppendorf AG