VIV102008SR3
SynVivo SynRAM 3D Inflammation model chip, IMN2 radial configuration
About This Item
Code UNSPSC :
60104506
Nomenclature NACRES :
NA.84
Produits recommandés
Conditionnement
pkg of 3 ea
Fabricant/nom de marque
SynVivo 102008-SR3
Température de stockage
room temp
Description générale
SynRAM Inflammation model chip - Barrier type: Pillars, Distance between pillars: 8 μm, Barrier height: 3 μm, Travel/barrier width: 50 μm, Outer Channel width: 200 μm, Device depth: 100 μm. This chip is used to create the SynRAM inflammation model in monoculture or co-culture configurations
Liaison
LInk to Publications application text