TMODS3111-0030
Nalgene® friction-fit tube closures
polypropylene, diam. 30 mm, push-fit, case of 20 ea
Synonyme(s) :
plastic centrifuge tube closures, polypropylene closures
About This Item
Code UNSPSC :
41121800
Nomenclature NACRES :
NB.43
Produits recommandés
Matériaux
polypropylene
Conditionnement
case of 20 ea
Fabricant/nom de marque
Nalgene DS3111-0030
Diamètre
30 mm
Catégories apparentées
Description générale
Easily remove Thermo Scientific Nalgene Friction-Fit Polypropylene Closures for Plastic Centrifuge Tubes from the centrifuge with a convenient pull tab. These autoclavable closures are designed to fit Nalgene round and conical centrifuge tubes. They may not be liquid-tight during high-speed centrifugation.
Informations légales
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries