TMO3410-08
Nalgene®PC Biotainer® Bottles and Carboys
clear polycarbonate, 10000 mL capacity, case of 2 ea
Synonyme(s) :
PETG bottle, bottle with handle, square bottle
About This Item
Produits recommandés
Matériaux
clear polycarbonate
Stérilité
sterile
Conditionnement
case of 2 ea
Fabricant/nom de marque
Nalgene 3410-08
Capacité
10000 mL
10000 mL capacity
Catégories apparentées
Description générale
Caractéristiques et avantages
- Durable, translucent PC ensures sample integrity
- White, silicone-lined, radiation stable polypropylene (PP) closures for leak resistance
- Graduations printed on bottles and molded into containers over 5L
- Individual customization options for fluid transfer
- Extensive validation documents are at your disposal
- Suitable for storage at temperatures from -100 °C to +100 °C