Principaux documents
TMO332189-0004
Nalgene® economy wide-mouth bottle
natural high-density polyethylene, capacity 125 mL, case of 500 ea
Synonyme(s) :
HDPE bottle, nalgene HDPE bottle
About This Item
Code UNSPSC :
41121800
Nomenclature NACRES :
NB.17
Matériaux
natural high-density polyethylene
Conditionnement
case of 500 ea
Fabricant/nom de marque
Nalgene 332189-0004
Capacité
125 mL
Description générale
Perform short-term general purpose laboratory and field applications with Thermo Scientific Nalgene Wide-Mouth HDPE Economy Bottles with Closure. These light-duty translucent bottles are a lightweight version of the original Nalgene bottle, bulk-packed for ultimate economy. Thinner-walled and more flexible than the Nalgene Lab Quality Bottles, they feature a wide mouth for easy filling and offer excellent chemical resistance to most acids, bases and alcohols.
Informations légales
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries