Principaux documents
TMO312760-0000
Nalgene® closure for dropper Bottle
natural polypropylene, diam. 16 mm, thread 15-415, case of 2000 ea
Synonyme(s) :
dropping bottle closure, dropping bottle seal
About This Item
Code UNSPSC :
41121800
Nomenclature NACRES :
NB.43
Matériaux
natural polypropylene
Conditionnement
case of 2000 ea
Fabricant/nom de marque
Nalgene 312760-0000
Diamètre
16 mm
Filetage
15-415
Description générale
Order Thermo Scientific Nalgene Closures for Dropper Bottles to complete the Nalgene Dropper Bottle System. Closures fit dropper bottles 312750 and 312751 and are available in multiple colors to help identify reagents.
Informations légales
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries