Principaux documents
TMO312018-1000
Nalgene® square graduated bottle
high-density polyethylene, bottle capacity 1000 mL, natural, case of 250 ea
Synonyme(s) :
break resistant bottle, chemcial resistant bottle
About This Item
Code UNSPSC :
41121800
Nomenclature NACRES :
NB.43
Matériaux
high-density polyethylene
natural
square
Conditionnement
case of 250 ea
Fabricant/nom de marque
Nalgene 312018-1000
Contenance du flacon
1000 mL
Catégories apparentées
Description générale
Save space with chemical- and break-resistant Thermo Scientific Nalgene Square HDPE Graduated Bottles with Closure. These translucent bottles provide visible liquid levels and molded-in graduations for easy measuring. Square bottles with polypropylene closures feature space-saving designs and drip-proof necks for convenient dispensing.
Informations légales
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries