Principaux documents
TMO2425-0502
Nalgene® narrow-mouth Right-to-Know wash bottle
low-density polyethylene, bottle volume 500 mL, ethyl alcohol, case of 24 ea
Synonyme(s) :
Nalgene bottle, plastic bottle
About This Item
Code UNSPSC :
41121800
Nomenclature NACRES :
NB.43
Matériaux
low-density polyethylene
narrow-mouth
Conditionnement
case of 24 ea
Fabricant/nom de marque
Nalgene 2425-0502
Volume du flacon
500 mL
Description générale
Meet regulatory labeling requirements for chemical containers with Thermo Scientific Nalgene Narrow-Mouth Right-to-Know LDPE Wash Bottles, which use standard, instantly recognizable symbols and colors. These low density polyethylene bottles are flexible for easy dispensing and translucent for viewing contents.
Informations légales
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries