Principaux documents
TMO232698
Nunc® RNAse/DNAse free sealing tape for 96-well plate
aluminum acrylate, non-sterile, case of 100 ea
Synonyme(s) :
sealing film, sealing foil
About This Item
Code UNSPSC :
41122102
Nomenclature NACRES :
NB.15
Stérilité
non-sterile
Conditionnement
case of 100 ea
Fabricant/nom de marque
Nunc 232698
Catégories apparentées
Description générale
Minimize evaporation and protect samples from contamination and spilling with Thermo Scientific Nunc Sealing Tapes. These adhesive seals are ideal for all assay applications. Nunc Life Science products offer reliability and reproducible results, and are backed by industry-leading quality standards and certifications.
Informations légales
Nunc is a registered trademark of Thermo Fisher Scientific or its subsidiaries