Principaux documents
TMO167574
Nunc® Edge 2.0 96-well Plate
Nunclon® Delta surface, polystyrene, sterile, without lid, clear, case of 50 ea
Synonyme(s) :
cell culture plate, cell growth plate
About This Item
Code UNSPSC :
41122102
Nomenclature NACRES :
NB.15
Matériaux
clear
polystyrene
Stérilité
sterile
Conditionnement
case of 50 ea
Fabricant/nom de marque
Nunc 167574
Description générale
Minimize plate evaporation and the risks associated with edge effect by using the uniquely engineered Thermo Scientific Nunc Edge 2.0 Plate. The surrounding bulit-in reservoirs, when filled with sterile water or media, act as evaporation barriers, yielding consistency across the entire 96-well plate. Enabling the use of all 96 wells in a plate allows researchers to plan experiments more efficiently with enhanced productivity and less waste for cell-based assays that require extended incubation.
Informations légales
Nunc is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Nunclon is a registered trademark of Thermo Fisher Scientific or its subsidiaries