Principaux documents
RA481500-EP
Rotor for Eppendorf® 5804/5804R & 5810/5810R Centrifuges
A-4-81, for 5810/5810R, aerosol-tight, with 4 × 500 mL rectangular buckets
About This Item
Code UNSPSC :
41103909
Caractéristiques
to hold 0.2 to 500 mL tubes and bottles
aerosol-tight
for 5810/5810R
medium to high-capacity swing-bucket rotor
with 4 × 500 mL rectangular buckets
Conditionnement
pack of 1 ea
Fabricant/nom de marque
Eppendorf® 022638602 (NA)
Eppendorf® 5810718007 (EU)
Paramètres
3,220 × g max. RCF
Technique(s)
centrifugation: suitable
Compatibilité
for use with centrifuge models 5804/5804R/5810/5810R
Description générale
A wide selections of rotors to fit the Eppendorf 5804 and 5810 centrifuges.
Informations légales
Eppendorf is a registered trademark of Eppendorf AG