Principaux documents
MTCV5920
MTC Bio Locking Rack for Cryogentic Vials
to hold, 5 x 10 cryovials
Synonyme(s) :
flask screw cap, vial holder, vial storage
About This Item
Code UNSPSC :
41122806
Nomenclature NACRES :
NB.12
Matériaux
polypropylene
Caractéristiques
to hold 5 x 10 cryovials
Conditionnement
pack of 4 ea
Fabricant/nom de marque
MTC Bio V5920
Description générale
Polypropylene rack for securely holding 50 "star foot" vials, 210 × 100 × 25mm, so that one hand is all that is needed to install and remove caps.
Informations légales
MTC is a trademark of MTC Bio