Principaux documents
MTCV5809B
MTC Bio Cap Inserts for Cryogenic Vials
blue
Synonyme(s) :
cryo storage identification, cryo vial marker, tube storage
About This Item
Code UNSPSC :
41122500
Nomenclature NACRES :
NB.11
Matériaux
blue
polypropylene
Conditionnement
pack of 500 ea
Fabricant/nom de marque
MTC Bio V5809-B
Description générale
Cap inserts for cryogenic vials, blue, 500/pk
Easily color-code your cryogenic vials for quick visual identification in the lab, for use at temperatures as low as -196°C.
Caractéristiques et avantages
  • For temperatures as low as -196°C
Informations légales
MTC is a trademark of MTC Bio