Principaux documents
MTCP6420
MTC Bio ProPette Manual Pipette Controller
with filter
Synonyme(s) :
pipettor
About This Item
Code UNSPSC :
41121500
Nomenclature NACRES :
NB.04
Conditionnement
pack of 1 ea
Fabricant/nom de marque
MTC Bio P6420
Catégories apparentées
Description générale
ProPette MPC is the precise and easy-to-use manual pipette controller that‘s always ready to use. After attaching a pipette, one squeeze of its large rubber filling bulb is all it takes to start pipetting. There are no wires, no tubing, and no battery to recharge. Begin aspirating by touching the EZ-Flow lever, which is designed to precisely control the filling speed based on how gently or firmly you press the lever. A built-in replaceable filter ensures an optimum rate and protects the controller from liquid penetration and overfill. A separate button provides a positive blow-out function when required.
Caractéristiques et avantages
  • For glass or plastic pipettes
  • Accommodates pipettes up to 100mL
  • Easy, single lever control
  • Autoclavable lower body
  • Fingertip speed control
Informations légales
MTC is a trademark of MTC Bio
ProPette is a trademark of MTC Bio