Principaux documents
MTCP52002U
MTC Bio ProPette LE Adjustable Pipettor
capacity 0.2-2.0 μL
Synonyme(s) :
pipette
About This Item
Code UNSPSC :
41121500
Nomenclature NACRES :
NB.15
Caractéristiques
adjustable
manual
no. of channels 1
Conditionnement
pack of 1 ea
Fabricant/nom de marque
MTC Bio P5200-2U
Capacité
0.2-2.0 μL
Catégories apparentées
Description générale
ProPette LE Single-Channel Pipettes provide maximum precision at virtually every conceivable volume. They feature a digital micrometer that is adjustable for precise volume control as well as a mechanical volume lock positioned at the top of the pipette that can be engaged or disengaged with one hand. These pipettors are lightweight and finely balanced for maximum ergonomics.
ProPette LE, 0.2 to 2µL
Caractéristiques et avantages
  • Adjustable with Roto-Lock
  • Light weight with reduced forces
  • Full volume range (0.1µL-10mL)
  • Superior accuracy and reproducibility
  • Outstanding user comfort
Informations légales
MTC is a trademark of MTC Bio
ProPette is a trademark of MTC Bio