Principaux documents
MTCP4300FT
MTC Bio Filtered Pipette Tips
capacity 300 μL, sterile, pkg of 960 ea (10 x racks 96ea)
Synonyme(s) :
conical tube, filter tip, pipetting tip
About This Item
Code UNSPSC :
41121601
Nomenclature NACRES :
NB.02
Matériaux
clear
polypropylene
Stérilité
sterile
Caractéristiques
DNase free
RNase free
Conditionnement
pkg of 960 ea (10 x racks 96ea)
Fabricant/nom de marque
MTC Bio P4300-FT
Capacité
300 μL
Volume
300 μL
Catégories apparentées
Description générale
Pipette Tips, Filtered, 300µL capacity, sterile, for 200µL and 300µL pipettes, 10 racks of 96 tips These tips are designed to fit MTC’s exclusive 300µL single channel ProPette™ LE as well as most popular multi-channel pipettes.
Caractéristiques et avantages
  • For 300µL pipettes
  • Also fits most 200µL pipettes
  • Filtered or non-filtered
  • Pre-sterilized
  • Autoclavable box-rack
Informations légales
MTC is a trademark of MTC Bio
ProPette is a trademark of MTC Bio