Principaux documents
MTCH68001MXL
MTC Bio Halo Lightweight Single Channel Pipettor
capacity 200-1250 μL
Synonyme(s) :
pipette
About This Item
Code UNSPSC :
41121500
Nomenclature NACRES :
NB.15
Caractéristiques
adjustable
manual
no. of channels 1
Conditionnement
pack of 1 ea
Fabricant/nom de marque
MTC Bio H6800-1MXL
Capacité
200-1250 μL
Catégories apparentées
Description générale
As one of the lightest pipettors ever introduced, the Halo Pipettor dramatically reduces the risk of repetitive stress injury and the general shoulder, arm, and hand fatigue associated with daily pipetting. Reduced actuation forces andan ergonomic design make Halo Pipettors feel balanced and natural in hand. Featuring a four-digit micrometer display for precise volume selection and a click-based adjustment wheel with DriftGuard protection, the Halo Pipettor is perfect for applications that demand exceptional accuracy. With a recessed ejector button, and cushioned, shock-absorbing stop, the ergonomics of tip ejection have been re-imagined to relieve stress on the thumb and joints.
Halo Single Channel Pipettor, lightweight, 10-100µL
Caractéristiques et avantages
  • 10-40% lighter than leading competitors
  • 4 Digit micrometer with DriftGuard protection
  • Exclusive 1250µL model for extended length tips
  • User-enabled calibration port
  • Compatible with universal tips
Informations légales
Halo is a trademark of MTC Bio
MTC is a trademark of MTC Bio