Principaux documents
MTCE4000VG1
MTC Bio SmartBlue UV Viewing Googles
pack of 1 ea
Synonyme(s) :
UV glasses
About This Item
Code UNSPSC :
46181802
Nomenclature NACRES :
NB.15
Conditionnement
pack of 1 ea
Fabricant/nom de marque
MTC Bio E4000-VG1
Description générale
SmartBlue UV viewing goggles, for blocking harmful UV rays from reaching eyes, 1/ea
Caractéristiques et avantages
  • Allow viewing of bands on the blue light transilluminator when the cover is not in place
  • Block the strong blue light and also block 100% of harmful UV light.
  • Optimized amber lenses block 99% of 465-475 nm blue light from LED transluminators
  • Allows clear visualization of green fluorescence
  • Protective case included
  • Adjustable arms for comfortable fit
Informations légales
MTC is a trademark of MTC Bio