Principaux documents
HW11200-02
Sigma-Aldrich
Lifting Bar, 4 Loop, ∅14" Body, Angled
About This Item
Code UNSPSC :
41116107
Nomenclature NACRES :
RA.01
Description générale
SAFC has qualified EZ BioPac, from ILC Dover as their preferred bulk powder package. Manufactured with Armorflex 114 film, the EzBiopac range is a fit for purpose, scalable and intuitive offer that enables the clean, safe and efficient delivery of bulk powders into bioprocess.
This is an accessory which can be used to assist with the handling of the EZ BioPac by ILC Dover filled with cell culture media.
Autres remarques
Low profile lifting bar for products DP4603, DP4604