Principaux documents
HS1040B
Earplug Dispenser Counter or Wall Mount
green (dispenser)
About This Item
Code UNSPSC :
41121800
Nomenclature NACRES :
NB.13
Matériaux
acrylic
green (dispenser)
Caractéristiques
holds Small laboratory consumables, items, and accessories
Conditionnement
pack of 1 ea
Fabricant/nom de marque
Heathrow Scientific HS1040B
L × l × H
155 mm × 155 mm × 170 mm
Description générale
The Translucent Earplug Dispenser provides easy access to safety accessories.
Caractéristiques et avantages
  • Place on counter or wall mount (screws included)
  • Holds approximately 200 pairs of foam earplugs