EPP0625-2110
Chart Recorder and Accessories for all Eppendorf® Freezer Models
paper, temp. range -50 — -100 °C, pack of 60
About This Item
Code UNSPSC :
41121800
Nomenclature NACRES :
NB.12
Produits recommandés
Matériaux
paper
Conditionnement
pack of 60
Fabricant/nom de marque
Eppendorf® P0625-2110
Paramètres
-50 — -100 °C temp. range
Catégories apparentées
Description générale
The chart recorder will continuously monitor and record freezer temperature. It is front mounted to your freezer for easy access and can be fitted to all freezer models.
Caractéristiques et avantages
- Includes chart recorder, RTD probe, keys, chart recorder paper, and chart recorder pens
- Will continuously monitor and record freezer temperature for data documentation
- Front mounted to your freezer for easy access and can be fitted to all freezer models
- Type I: Battery powered, will provide a minimum of eighteen months of operation
- User configurable built in high and low temperature alarms
- Can be factory installed with your freezer order at no additional charge or retrofitted on location
- Selectable temperature range (0 to -50 C and -50 to -100 C) and rotation speed
Informations légales
Eppendorf is a registered trademark of Eppendorf AG