EPM1335-0600
12 Port Gassing Manifold
for Eppendorf Brunswick Innova 42/42R
About This Item
Code UNSPSC :
41104405
Nomenclature NACRES :
NB.73
Produits recommandés
Fabricant/nom de marque
Eppendorf® M1335-0600 (Global)
Catégories apparentées
Description générale
The manifold delivers gas into the chamber and ultimately into the flasks themselves via twelve ports. You may elect to use splitters after the manifold to increase the number of ports for your application. Gassing manifolds are a factory fitted option and need specialist installation.
Informations légales
Eppendorf is a registered trademark of Eppendorf AG