Principaux documents
EPM1194-9903
Platform for Eppendorf® New Brunswick Innova® 2100/40/40R/42/42R & Excella® E24/E24R Shakers
for 50 mL Erlenmeyer flasks, non-Interchangeable, phenolic resin
About This Item
Code UNSPSC :
41103816
Nomenclature NACRES :
NB.73
Caractéristiques
64 x 50 mL Erlenmeyer flasks
for 50 mL Erlenmeyer flasks
non-Interchangeable
phenolic resin
Fabricant/nom de marque
Eppendorf® M1194-9903 (Global)
L × l
45.7 cm × 45.7 cm
Catégories apparentées
Description générale
Platforms designed for the Eppendorf New Brunswick Innova 2100/40/40R/42/42R & Excella E24/E24R shakers to ensure a safe and perfect fit.
Informations légales
Eppendorf is a registered trademark of Eppendorf AG
Excella is a registered trademark of Eppendorf, Inc. 
Innova is a registered trademark of Eppendorf, Inc. 
New Brunswick is a trademark of Eppendorf AG