Principaux documents
EPM1190-9921
Platform Clamps, for Eppendorf® Shakers
for Eppendorf® New Brunswick Innova, I-Series and Excella shakers, starter kit
About This Item
Code UNSPSC :
41122413
Caractéristiques
for Eppendorf® New Brunswick Innova, I-Series and Excella shakers
includes 4x 250 mL, 4x 500 mL, 2x 1 L, and 2x 2 L flask clamps
starter kit
Fabricant/nom de marque
Eppendorf® M1190-9921 (NA)
Catégories apparentées
Description générale
Flask clamps are of a single piece stainless-steel construction (not spot welded or riveted) for added strength and safety. Clamps 2 L and above come with an extra girdle for added security when agitating flasks at high speeds. Flask clamps come complete with required screws, and are fitted with coiled wire springs for added stability at elevated speeds.
Informations légales
Eppendorf is a registered trademark of Eppendorf AG