EPF740520025
Eppendorf® CryoCube® F740hi
capacity 740 L, touchscreen interface, cooling, 3, AC/DC input 120 - 208 V, 60 Hz
Synonyme(s) :
CryoCube® F740hi, CryoCube® F740hi-ULT Freezer, Eppendorf® CryoCube® F740hi
About This Item
Code UNSPSC :
41103034
Produits recommandés
Caractéristiques
cooling 3
touchscreen interface
Capacité
740 L
Description générale
The Eppendorf CryoCube F740 series is one of the newest additions to the Eppendorf ultra-low temperature freezer family (also called ultra-low temperature freezer, -80C). The CryoCube F740 series of ultra-low temperature freezers combines the traditionally high quality of our previous ultra-low temperature freezers with a higher capacity to safely store more of your samples while optimizing energy consumption. By incorporating the PhysioCare Concept into the development, the CryoCube F740 is lighter and more comfortable to use, supports ergonomics in the workplace and enables an optimal workflow in the laboratory.
Application
Cryopreservation, Cryostorage
Informations légales
CryoCube is a registered trademark of Eppendorf AG
Eppendorf is a registered trademark of Eppendorf AG