Principaux documents
EP952010051
Eppendorf® Uvette® 220 nm – 1,600 nm
PCR clean, capacity 50 -– 2,000  μL, pack of 80 ea
About This Item
Code UNSPSC :
41121813
Matériaux
(UV-transparent plastic)
Caractéristiques
PCR clean
Conditionnement
pack of 80 ea
Fabricant/nom de marque
Eppendorf® 952010051
Capacité
50 -– 2,000  μL
Description générale
UVette® 220 nm – 1,600 nm, original Eppendorf plastic cuvette, individually packed, (including RNase-free, DNase-free) and protein-free, PCR clean, 50 – 2,000 µL, 80 pcs.
  • individually packaged
  • PCR-clean and protein-free quality
  • UV transparent plastic, free of fluoropolymers and other halogenated hydrocarbons
  • Two built-in optical path lengths in a single cuvette - just turn the UVette 90° to change from 10 mm and 2 mm
  • Volume markings for 500 µL and 1,000 µL
  • Individually packaged and certified PCR clean and protein-free quality for sensitive (e.g. RNA) and valuable samples
  • Deep-lying optical window prevents scratches
  • Tapered cuvette base for optimal filling, frosted grip for labeling
  • Ideal for use in the Eppendorf BioPhotometer and Eppendorf BioSpectrometer, adapters available for use with other common spectrophotometers
  • Routine pack for convenient access to each UVette and safe storage in a reclosable box
Caractéristiques et avantages
  • Individually packaged
  • PCR-clean and protein-free quality
  • UV transparent plastic, free of fluoropolymers and other halogenated hydrocarbons
  • Two built-in optical path lengths in a single cuvette - just turn the UVette 90° to change from 10 mm and 2 mm
  • Volume markings for 500 µL and 1,000 µL
  • Individually packaged and certified PCR clean and protein-free quality for sensitive (e.g. RNA) and valuable samples
  • Deep-lying optical window prevents scratches
  • Tapered cuvette base for optimal filling, frosted grip for labeling
  • Ideal for use in the Eppendorf BioPhotometer and Eppendorf BioSpectrometer, adapters available for use with other common spectrophotometers
  • Routine pack for convenient access to each UVette and safe storage in a reclosable box
Informations légales
Eppendorf is a registered trademark of Eppendorf AG
UVette is a registered trademark of Eppendorf AG