Principaux documents
EP951010049
Eppendorf® Rotor Accessories
work tray frame, for 96 x 0.2 mL PCR tubes, pack of 5
About This Item
Code UNSPSC :
41103911
Caractéristiques
Holds 0.2 mL tubes
for 96 x 0.2 mL PCR tubes
Conditionnement
pack of 5
Fabricant/nom de marque
Eppendorf® 951010049 (US)
Technique(s)
PCR: suitable
centrifugation: suitable
Compatibilité
for use with Rotor A-2-VC
Informations légales
Eppendorf is a registered trademark of Eppendorf AG