Principaux documents
EP6731011025
Eppendorf® CellXpert® C170i CO2 Incubator
Oxygen control, inner door non-segmented, handle left side, AC/DC input 120 V, US 2-pin plug (US/JP/South America)
Synonyme(s) :
Heaters
About This Item
Code UNSPSC :
41104405
Conditionnement
pack of 1 ea
Fabricant/nom de marque
Eppendorf® 6731011025
Description générale
CellXpert® C170i, non-segmented inner door, handle left side, 1 – 20 % oxygen control, 100 – 127 V/50 – 60 Hz (US/JP/South America)
Caractéristiques et avantages
  • 1 – 20 % oxygen control
  • handle left side
  • non-segmented inner door
  • 180° HTD (standard with C170i)
  • including water tray and four shelves (stainless steel)
  • 100 – 127 V, 50 – 60 Hz
Informations légales
CellXpert is a registered trademark of Eppendorf AG
Eppendorf is a registered trademark of Eppendorf AG