Principaux documents
EP5910706006
Eppendorf® Adapter
for use with for 400 mL bottle, for S-4x400 rotor, set of 2 pieces
About This Item
Code UNSPSC :
41121815
Nomenclature NACRES :
NB.82
Conditionnement
set of 2 pieces
Fabricant/nom de marque
Eppendorf® 5910706006
Eppendorf®
Compatibilité
for use with for 400 mL bottle, for S-4x400 rotor
Informations légales
Eppendorf is a registered trademark of Eppendorf AG