Principaux documents
EP5820761008
Lid for Eppendorf(R) FA-45-48-11 Rotor
aerosol-tight, for 5804/5804R & 5810/5810R, aluminum
About This Item
Code UNSPSC :
41103913
Nomenclature NACRES :
NB.82
Matériaux
aluminum
Caractéristiques
aerosol-tight
Fabricant/nom de marque
Eppendorf® 5820761008 (Global)
Technique(s)
centrifugation: suitable
Compatibilité
for use with 5804
for use with 5804 R
for use with 5810 R
for use with 5810
Informations légales
Eppendorf is a registered trademark of Eppendorf AG