Principaux documents
EP5391000010
Eppendorf(R) HeatSealer
S100
About This Item
Code UNSPSC :
41102400
Entrée CA/CC
100 - 230 V, 50/60 Hz, US 2-pin plug
Caractéristiques
CE compliant
CSA approved
fixed Sealing, temperature
seals 96 and 384 well plates
Fabricant/nom de marque
Eppendorf® 5391000010 (US)
Catégories apparentées
Description générale
The Eppendorf HeatSealer Family offers you reliable and effective Sealing, of a variety of plates. You can choose from a more compact (S100) and a more flexible (S200) instrument according to your Sealing, needs.
Caractéristiques et avantages
Hermetic Sealing, of multiwell plates
Eliminates evaporation in PCR
Seals 96- and 384-well plates
Integrated thermostat prevents overheating
Improved mechanics for even easier Sealing,
PCR Plate Adapter included
Informations légales
Eppendorf is a registered trademark of Eppendorf AG