Principaux documents
EP022822161
Rotor for Eppendorf® Vacufuge® Plus Concentrator
F-40-18-19, non aerosol tight, (8 x 10.0 mL flat-bottom tubes)
About This Item
Code UNSPSC :
41103909
Caractéristiques
to hold 8 x 10 mL flat-bottom tubes
to hold 8 x 19x66 mm
fixed angle rotor
non aerosol tight
Conditionnement
pack of 1 ea
Fabricant/nom de marque
Eppendorf® 022822161 (NA)
Technique(s)
centrifugation: suitable
Capacité
(8 x 10.0 mL flat-bottom tubes)
Compatibilité
for use with centrifuge models Vacufuge Plus Concentrator
Description générale
Selection of rotors for the Eppendorf Vacufuge Plus.
Informations légales
Eppendorf is a registered trademark of Eppendorf AG
Vacufuge is a registered trademark of Eppendorf AG