Principaux documents
EP022639528
Adapter for Eppendorf® A-8-17 Rotor
holds 1 x 15 mL conical tube, pack of 8 ea
About This Item
Code UNSPSC :
41103911
Matériaux
POM
Caractéristiques
holds 1 x 15 mL conical tube
Conditionnement
pack of 8 ea
Fabricant/nom de marque
Eppendorf® 022639528 (NA)
Technique(s)
centrifugation: suitable
Compatibilité
for use with Rotor A-8-17
Description générale
Adapter, for 1 conical tube 15 mL, for Rotor A-8-17, 8 pcs.
Informations légales
Eppendorf is a registered trademark of Eppendorf AG