Principaux documents
EP0030081234
Eppendorf® BioBased ep Dualfilter T.I.P.S. SealMax
greener alternative
volume 0.5—20 μL, sterile, Biopur, L -46 mm, pkg of 960 ea (10 trays x 96 tips)
About This Item
Code UNSPSC :
41121600
Stérilité
sterile
Caractéristiques
DNase free
RNase free
graduations
Biopur
Conditionnement
pkg of 960 ea (10 trays x 96 tips)
Fabricant/nom de marque
Eppendorf® 0030081234
Caractéristiques du produit alternatif plus écologique
Use of Renewable Feedstocks
Learn more about the Principles of Green Chemistry.
sustainability
Greener Alternative Product
Technique(s)
DNA purification: suitable
PCR: suitable
Longueur
-46 mm
Volume
0.5—20 μL
Adéquation
suitable for PCR
suitable for microbiology
suitable for molecular biology
suitable for nucleic acid purification
Autre catégorie plus écologique
Description générale
We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product aligns with ″Use of Renewable Feedstocks″. Click here for more information.
Application
Molecular Biology, DNA purification, PCR, Microbiology, Cell Biology, Medical Research, Agriculture Research, Environmental Monitoring, Forensic Analysis
Caractéristiques et avantages
  • Improve your laboratory carbonfootprint and switch to epT.I.P.S. BioBased in the newEppendorf Reload variants. These pipette tips are produced from second-generationbiobased polymer with 90% of the feedstock attributed to renewable resources (i.e. waste and residues from vegetable oilrefining or waste cooking oil), avoiding the use of fossil fuel-based petroleum
  • Reduce your laboratory waste volume by useing Reloads instead of disposable Racks
  • Eppendorf production facility is ISCC PLUS(International Sustainability & Carbon Certification) certified, confirmingthe raw materials used at the facility are “bio-circular”. Enables traceabilityalong the entire supply chain
  • Pipette tips with ACT (Accountability, Consistency, and Transparency) Label- an independent environmental impact factor assessment through non-profit MyGreen Lab
  • Material, quality and purity certified
  • Get excellent sample integrity as no slip agents, plasticizers and biocides are used
  • Independent laboratory – Lot-specific purity certificates can be downloaded from Eppendorf website
  • General quality certificates in compliance with standards and general Eppendorf quality items
  • Available in volumes from 10 µL to 1,250 µL Long in Reloads for use with epT.I.P.S. Box 2.0
  • Available as epT.I.P.S. BioBased Biopur, ep Dualfilter T.I.P.S. BioBased PCR clean and sterile, ep Dualfilter T.I.P.S. SealMax BioBased Biopur
Informations légales
Eppendorf is a registered trademark of Eppendorf AG