Principaux documents
CLS766113
Pyrex® pennyhead stopper
stopper joint: 13
Synonyme(s) :
glass stopper, glass stoppers, stoppers, Corning® 7660
About This Item
Code UNSPSC :
41122500
Matériaux
glass
Joint
joint: stopper13
Fabricant/nom de marque
Corning 7661-13
Description générale
Ground zone length indicates its greatest diameter (mm).
Informations légales
Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.